Alfa Aesar™ 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine, 95%

Manufacturer: Alfa Aesar™ H2691103

Quantity 1g

 View more versions of this product

Catalog No. AAH2691103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 885952-23-8
    Molecular Formula C15H16BrN3O2
    Formula Weight 350.21
    MDL Number MFCD06798240
    Chemical Name or Material 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine

    Specifications

    Formula Weight 350.21
    Molecular Formula C15H16BrN3O2
    MDL Number MFCD06798240
    CAS 885952-23-8
    Melting Point 114° to 116°C
    Quantity 1g
    Chemical Name or Material 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit