Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldicarboxylic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
787-70-2
C14H10O4
242.23
MFCD00002554
NEQFBGHQPUXOFH-UHFFFAOYSA-N
biphenyl-4,4'-dicarboxylic acid,4,4'-biphenyldicarboxylic acid,1,1'-biphenyl-4,4'-dicarboxylic acid,4-4-carboxyphenyl benzoic acid,4,4'-bibenzoic acid,4,4'-diphenic acid,4,4'-dicarboxybiphenyl,p,p'-diphenic acid,usaf do-69,bibenzoic acid
13084
4-(4-carboxyphenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O
787-70-2
C14H10O4
242.23
MFCD00002554
NEQFBGHQPUXOFH-UHFFFAOYSA-N
biphenyl-4,4'-dicarboxylic acid,4,4'-biphenyldicarboxylic acid,1,1'-biphenyl-4,4'-dicarboxylic acid,4-4-carboxyphenyl benzoic acid,4,4'-bibenzoic acid,4,4'-diphenic acid,4,4'-dicarboxybiphenyl,p,p'-diphenic acid,usaf do-69,bibenzoic acid
13084
4-(4-carboxyphenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O
Color White to Beige
Quantity 1g
Melting Point 300.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (C6H4COOH)2
Formula Weight 242.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4, 4'-Biphenyldicarboxylic acid
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 212-328-3