Alfa Aesar™ 4,4'-Bipyridine, 98%

Manufacturer: Alfa Aesar™ A1018722

Quantity 100g

 View more versions of this product

Catalog No. AAA1018722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 553-26-4
    Molecular Formula C10H8N2
    Formula Weight 156.19
    MDL Number MFCD00006416
    UN Number UN2811
    Beilstein 113176
    Merck Index 14,3348
    Synonym 4,4'-Bipyridyl; 4,4'-Dipyridyl
    Chemical Name or Material 4,4'-Bipyridine

    Specifications

    Formula Weight 156.19
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 553-26-4
    Merck Index 14,3348
    Quantity 100g
    Health Hazard 1 Danger
    TSCA TSCA
    Molecular Formula C10H8N2
    MDL Number MFCD00006416
    UN Number UN2811
    Boiling Point 304° to 305°C
    Health Hazard 2 GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Beilstein 113176
    Melting Point 111° to 114°C
    Synonym 4,4'-Bipyridyl; 4,4'-Dipyridyl
    RTECS Number DW1760000
    Chemical Name or Material 4,4'-Bipyridine
    Assay 98%

    Description

    Applications
    4,4 ft.-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N ft.-dimethyl-4,4 ft.-bipyridinium.

    Solubility
    Soluble in ethanol, ether. Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents and strong acids.

    GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.

    GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit