Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bipyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7080603
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
553-26-4
C10H8N2
156.19
MFCD00006416
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
11107
CHEBI:30985
4-pyridin-4-ylpyridine
C1=CC(=CC=N1)C1=CC=NC=C1
This item is not returnable. View return policy
553-26-4
C10H8N2
156.19
MFCD00006416
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
11107
CHEBI:30985
4-pyridin-4-ylpyridine
C1=CC(=CC=N1)C1=CC=NC=C1

4,4'-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N'-dimethyl-4,4'-bipyridinium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N′-dimethyl-4,4′-bipyridinium.

Solubility
Soluble in ethanol, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 111°C to 114°C
Boiling Point 304°C to 305°C
Quantity 5 g
UN Number UN2811
Beilstein 113176
Sensitivity Hygroscopic
Merck Index 14,3348
Solubility Information Soluble in ethanol,ether. Insoluble in water.
Formula Weight 156.19
Percent Purity 98%
Chemical Name or Material 4,4'-Bipyridine
Show More Show Less
Hazard Category H301+H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-036-3
RTECSNumber DW1760000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit