Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Chlorobenzoyl)piperidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
53220-41-0
C12H14ClNO
223.7
MFCD04113538
IYGWDOXHCPQXKN-UHFFFAOYSA-N
4-4-chlorobenzoyl piperidine,4-chlorophenyl piperidin-4-yl methanone,4-4-chlorophenyl carbonyl piperidine,4-chlorophenyl-4-piperidinyl methanone,4-chlorophenyl-piperidin-4-yl-methanone,maybridge1_003174,4-chlorophenyl 4-piperidinyl methanonehydrochloride,4-4-chlorobenzoyl-piperidine,4-chlorophenyl 4-piperidyl ketone,4-chlorophenyl piperidin-4-ylmethanone
2777115
(4-chlorophenyl)-piperidin-4-ylmethanone
C1CNCCC1C(=O)C2=CC=C(C=C2)Cl
53220-41-0
C12H14ClNO
223.7
MFCD04113538
IYGWDOXHCPQXKN-UHFFFAOYSA-N
4-4-chlorobenzoyl piperidine,4-chlorophenyl piperidin-4-yl methanone,4-4-chlorophenyl carbonyl piperidine,4-chlorophenyl-4-piperidinyl methanone,4-chlorophenyl-piperidin-4-yl-methanone,maybridge1_003174,4-chlorophenyl 4-piperidinyl methanonehydrochloride,4-4-chlorobenzoyl-piperidine,4-chlorophenyl 4-piperidyl ketone,4-chlorophenyl piperidin-4-ylmethanone
2777115
(4-chlorophenyl)-piperidin-4-ylmethanone
C1CNCCC1C(=O)C2=CC=C(C=C2)Cl
Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Melting Point 60°C to 63°C
Quantity 5g
Color Yellow
Beilstein 158791
Solubility Information Slightly soluble in water.
Formula Weight 223.7
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures