Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Chlorobenzoyl)pyridine, 99+%

Click to view available options
Quantity:
1g
5g
25g
14548-48-2
C12H8ClNO
217.652
MFCD00006431
YMTFKKLFLLAFNI-UHFFFAOYSA-N
4-4-chlorobenzoyl pyridine,4-chlorophenyl pyridin-4-yl methanone,4-4'-chlorobenzoyl pyridine,4-chlorophenyl-pyridin-4-ylmethanone,methanone, 4-chlorophenyl-4-pyridinyl,acmc-1bu0c,4-4-chloro-benzoyl pyridine,4-chlorophenyl 4-pyridyl ketone,4-chlorophenyl pyridin-4-ylmethanone,4-chlorophenyl-4-pyridyl methanone
98503
(4-chlorophenyl)-pyridin-4-ylmethanone
C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl
14548-48-2
C12H8ClNO
217.652
MFCD00006431
YMTFKKLFLLAFNI-UHFFFAOYSA-N
4-4-chlorobenzoyl pyridine,4-chlorophenyl pyridin-4-yl methanone,4-4'-chlorobenzoyl pyridine,4-chlorophenyl-pyridin-4-ylmethanone,methanone, 4-chlorophenyl-4-pyridinyl,acmc-1bu0c,4-4-chloro-benzoyl pyridine,4-chlorophenyl 4-pyridyl ketone,4-chlorophenyl pyridin-4-ylmethanone,4-chlorophenyl-4-pyridyl methanone
98503
(4-chlorophenyl)-pyridin-4-ylmethanone
C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl
Applications
4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 107°C to 109°C
Quantity 25g
Formula Weight 217.66
Percent Purity ≥99%
Chemical Name or Material 4-(4-Chlorobenzoyl)pyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning