Alfa Aesar™ 4,4'-Di-tert-butyldiphenylmethane, 99%

Manufacturer: Alfa Aesar™ B2182030

Quantity 250g

 View more versions of this product

Catalog No. AAB2182030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19099-48-0
    Molecular Formula C21H28
    Formula Weight 280.45
    MDL Number MFCD00043733
    Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane

    Specifications

    Formula Weight 280.45
    Melting Point 71° to 73°C
    Quantity 250g
    Molecular Formula C21H28
    MDL Number MFCD00043733
    CAS 19099-48-0
    Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane
    Assay 99%

    Description

    Applications
    It is employed as a intermediate for pharmaceuticals.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit