Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Di-tert-butyldiphenylmethane, 99%

Click to view available options
Quantity:
10g
50g
250g
19099-48-0
C21H28
280.455
MFCD00043733
KRMBVBPNWUBFBC-UHFFFAOYSA-N
4,4'-di-tert-butyldiphenylmethane,1-tert-butyl-4-4-tert-butylphenyl methyl benzene,bis 4-tert-butyl phenyl methane,4,4<<-di-tert-butyldiphenylmethane,1,1'-methylenebis 4-tert-butylbenzene,1-tert-butyl-4-4-tert-butylbenzyl benzene,1-tert-butyl-4-4-tert-butyl phenyl methyl benzene
1652739
1-tert-butyl-4-[(4-tert-butylphenyl)methyl]benzene
CC(C)(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)C(C)(C)C
19099-48-0
C21H28
280.455
MFCD00043733
KRMBVBPNWUBFBC-UHFFFAOYSA-N
4,4'-di-tert-butyldiphenylmethane,1-tert-butyl-4-4-tert-butylphenyl methyl benzene,bis 4-tert-butyl phenyl methane,4,4<<-di-tert-butyldiphenylmethane,1,1'-methylenebis 4-tert-butylbenzene,1-tert-butyl-4-4-tert-butylbenzyl benzene,1-tert-butyl-4-4-tert-butyl phenyl methyl benzene
1652739
1-tert-butyl-4-[(4-tert-butylphenyl)methyl]benzene
CC(C)(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)C(C)(C)C
Applications
It is employed as a intermediate for pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 10g
Melting Point 71°C to 73°C
Formula Weight 280.45
Percent Purity 99%
Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane