Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromobiphenyl, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
92-86-4
C12H8Br2
312.00
MFCD00000101
HQJQYILBCQPYBI-UHFFFAOYSA-N
4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
7110
1-bromo-4-(4-bromophenyl)benzene
BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
92-86-4
C12H8Br2
312.00
MFCD00000101
HQJQYILBCQPYBI-UHFFFAOYSA-N
4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
7110
1-bromo-4-(4-bromophenyl)benzene
BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Flash Point >100°C
Boiling Point 355°C to 360°C
Packaging Plastic bottle
Quantity 500g
Melting Point 165°C to 169°C
Color Undesignated
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula BrC6H4C6H4Br
Beilstein 05, 580
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol;,soluble in benzene
Formula Weight 312
Physical Form Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material 4, 4'-Dibromobiphenyl, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-198-6

TSCA : TSCA