Alfa Aesar™ 4,4'-Dibromooctafluorobiphenyl, 99%

Manufacturer: Alfa Aesar™ B2512606

Quantity 5g

 View more versions of this product

Catalog No. AAB2512606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10386-84-2
    Molecular Formula C12Br2F8
    Formula Weight 455.94
    MDL Number MFCD00000310
    UN Number UN3152
    Chemical Name or Material 4,4'-Dibromooctafluorobiphenyl

    Specifications

    Formula Weight 455.94
    Quantity 5g
    MDL Number MFCD00000310
    UN Number UN3152
    Melting Point 113° to 115°C
    Molecular Formula C12Br2F8
    CAS 10386-84-2
    DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
    Chemical Name or Material 4,4'-Dibromooctafluorobiphenyl
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit