Alfa Aesar™ 4,4-Diethylamino-2-butyn-1-ol, 98%

Manufacturer: Alfa Aesar™ H5350106

Quantity 5g

 View more versions of this product

Catalog No. AAH5350106

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10575-25-4
    Molecular Formula C8H15NO
    Linear Formula (CH3CH2)2NCH2C≡CCH2OH
    Formula Weight 141.21
    MDL Number MFCD00671356
    Chemical Name or Material 4,4-Diethylamino-2-butyn-1-ol

    Specifications

    Formula Weight 141.21
    Flash Point 104°C (219°F)
    Molecular Formula C8H15NO
    Linear Formula (CH3CH2)2NCH2C≡CCH2OH
    MDL Number MFCD00671356
    CAS 10575-25-4
    Boiling Point 231° to 232°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Chemical Name or Material 4,4-Diethylamino-2-butyn-1-ol
    Assay 98%

    Description

    Applications
    4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

    Solubility
    Soluble in chloroform and ethyl acetate. Soluble in water 11 g/L @25°C.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit