Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorobenzhydrol, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
365-24-2
C13H10F2O
220.219
MFCD00000357
WCTZPQWLFWZYJE-UHFFFAOYSA-N
4,4'-difluorobenzhydrol,bis 4-fluorophenyl methanol,bis-4-fluorophenyl-methanol,4,4'-difluorobenzhydryl alcohol,bis 4-fluorophenyl methan-1-ol,4,4'-diflurobenzylmethanol,pubchem14058,4,4'-diflourobenzhydrol,4,4'-difluoro benzhydrol,4,4'-difluoro-benzhydrol
123049
bis(4-fluorophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)O)F
365-24-2
C13H10F2O
220.219
MFCD00000357
WCTZPQWLFWZYJE-UHFFFAOYSA-N
4,4'-difluorobenzhydrol,bis 4-fluorophenyl methanol,bis-4-fluorophenyl-methanol,4,4'-difluorobenzhydryl alcohol,bis 4-fluorophenyl methan-1-ol,4,4'-diflurobenzylmethanol,pubchem14058,4,4'-diflourobenzhydrol,4,4'-difluoro benzhydrol,4,4'-difluoro-benzhydrol
123049
bis(4-fluorophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)O)F
Quantity 25g
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 45°C to 49°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (FC6H4)2CHOH
Beilstein 06, III, 3375
Formula Weight 220.22
Physical Form Crystalline Flakes or Powder
Percent Purity ≥97.5%
Chemical Name or Material 4, 4'-Difluorobenzhydrol, 98%
Show More Show Less