Alfa Aesar™ 4,4'-Difluorobenzhydryl chloride, 98%

Manufacturer: Alfa Aesar™ A1122618

Quantity 50g

 View more versions of this product

Catalog No. AAA1122618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27064-94-4
    Molecular Formula C13H9ClF2
    Formula Weight 238.66
    MDL Number MFCD00044329
    UN Number UN1760
    Beilstein 2052680
    Synonym Chlorobis(4-fluorophenyl)methane
    Chemical Name or Material 4,4'-Difluorobenzhydryl chloride

    Specifications

    Formula Weight 238.66
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 27064-94-4
    Quantity 50g
    MDL Number MFCD00044329
    UN Number UN1760
    Health Hazard 1 Danger
    Boiling Point 126° to 128°C (1mmHg)
    Molecular Formula C13H9ClF2
    Synonym Chlorobis(4-fluorophenyl)methane
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flash Point >110°C (230°F)
    Beilstein 2052680
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Chemical Name or Material 4,4'-Difluorobenzhydryl chloride
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit