Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorochalcone, 98%, predominantly trans, ACROS Organics™

Click to view available options
Quantity:
5g
2805-56-3
C15H10F2O
244.241
MFCD00219906
AZBVDMNDUOURGI-XCVCLJGOSA-N
4,4'-difluorochalcone,e-1,3-bis 4-fluorophenyl prop-2-en-1-one,2e-1,3-bis 4-fluorophenyl prop-2-en-1-one,1,3-bis 4-fluorophenyl prop-2-en-1-one,trans-4,4'-difluorochalcone,2-propen-1-one,1,3-bis 4-fluorophenyl,4,4'-difluoro chalcone,4,4/'-difluorochalcone,e-4,4'-difluorochalcone,azbvdmnduourgi-xcvcljgosa
5286094
(E)-1,3-bis(4-fluorophenyl)prop-2-en-1-one
C1=CC(=CC=C1C=CC(=O)C2=CC=C(C=C2)F)F
2805-56-3
C15H10F2O
244.241
MFCD00219906
AZBVDMNDUOURGI-XCVCLJGOSA-N
4,4'-difluorochalcone,e-1,3-bis 4-fluorophenyl prop-2-en-1-one,2e-1,3-bis 4-fluorophenyl prop-2-en-1-one,1,3-bis 4-fluorophenyl prop-2-en-1-one,trans-4,4'-difluorochalcone,2-propen-1-one,1,3-bis 4-fluorophenyl,4,4'-difluoro chalcone,4,4/'-difluorochalcone,e-4,4'-difluorochalcone,azbvdmnduourgi-xcvcljgosa
5286094
(E)-1,3-bis(4-fluorophenyl)prop-2-en-1-one
C1=CC(=CC=C1C=CC(=O)C2=CC=C(C=C2)F)F
Color White to Yellow
Melting Point 113°C to 118°C
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 244.06
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4, 4'-Difluorochalcone, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.