Learn More
Medchemexpress LLC 4,4'-dihydroxybenzophenone | 611-99-4 | MFCD00002358 | 99.8% | 214.22 g·mol⁻¹ | C13H10O3 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW014226500G
Encompass_Preferred
4,4'-Dihydroxybenzophenone (CAS 611-99-4) is a phenolic ultraviolet absorber and chemical intermediate used in research and synthesis. It is a crystalline organic compound (C13H10O3, MW 214.22 g·mol⁻¹) commonly employed as a UV filter and as a building block in pharmaceutical syntheses. Manufacturer data report high purity and environmental toxicity notes that should be reviewed in the safety data sheet.
  • Phenolic ultraviolet absorber for UV-filter applications.
  • Chemical intermediate for synthesis of pharmaceutical compounds.
  • High reported purity, suitable for research-grade syntheses.
  • Well characterized: CAS 611-99-4, formula C13H10O3, molecular weight 214.22 g·mol⁻¹.
  • Documentation includes toxicity and purity details; consult the safety data sheet before use.
Catalog No. 50-003-04189