Alfa Aesar™ 4,4'-Diiodobiphenyl, 99%

Manufacturer: Alfa Aesar™ B2197014

Quantity 25g

 View more versions of this product

Catalog No. AAB2197014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3001-15-8
    Molecular Formula C12H8I2
    Formula Weight 406
    MDL Number MFCD00001057
    UN Number UN3152
    Chemical Name or Material 4,4'-Diiodobiphenyl

    Specifications

    Formula Weight 406
    TSCA TSCA
    Melting Point 205° to 211°C
    Quantity 25g
    MDL Number MFCD00001057
    CAS 3001-15-8
    Molecular Formula C12H8I2
    UN Number UN3152
    DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
    Chemical Name or Material 4,4'-Diiodobiphenyl
    Assay 99%

    Description

    Applications
    4,4 ft.-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4 ft.-diiodobipheny enhanced the expression of the luciferase gene.

    Solubility
    Soluble in hot Toluene(very faint turbidity).

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit