Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diiodobiphenyl 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3001-15-8
C12H8I2
406
MFCD00001057
GPYDMVZCPRONLW-UHFFFAOYSA-N
4,4'-diiodobiphenyl,4,4'-diiodo-1,1'-biphenyl,4,4'-diiododiphenyl,1,1'-biphenyl, 4,4'-diiodo,1-iodo-4-4-iodophenyl benzene,4,4-diiodobiphenyl,4-4'-iodophenyl iodobenzene,4,4;-diiodobiphenyl,4,4'-diiododophenyl, pract,4-iodo-1-4-iodophenyl benzene
76348
1-iodo-4-(4-iodophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)I)I
3001-15-8
C12H8I2
406
MFCD00001057
GPYDMVZCPRONLW-UHFFFAOYSA-N
4,4'-diiodobiphenyl,4,4'-diiodo-1,1'-biphenyl,4,4'-diiododiphenyl,1,1'-biphenyl, 4,4'-diiodo,1-iodo-4-4-iodophenyl benzene,4,4-diiodobiphenyl,4-4'-iodophenyl iodobenzene,4,4;-diiodobiphenyl,4,4'-diiododophenyl, pract,4-iodo-1-4-iodophenyl benzene
76348
1-iodo-4-(4-iodophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)I)I
Melting Point 201°C to 204°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula IC6H4C6H4I
Beilstein 05, 581
Formula Weight 406
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 4, 4'-Diiodobiphenyl
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning