Learn More
4,4'-Dimethoxybenzophenone, 98+%, Thermo Scientific Chemicals
Description
4,4'-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,4′-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is insoluble in water, soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong reducing agents.
Specifications
Melting Point | 142°C to 146°C |
Quantity | 25 g |
Beilstein | 1878026 |
Solubility Information | It is insoluble in water,soluble in ethanol. |
Formula Weight | 242.27 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,4'-Dimethoxybenzophenone |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 202-028-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.