Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxytrityl Chloride, 98%, Thermo Scientific Chemicals

Catalog Number AC146020250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
40615-36-9
C21H19ClO2
338.83
MFCD00008409
JBWYRBLDOOOJEU-UHFFFAOYSA-N
4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
96831
1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
40615-36-9
C21H19ClO2
338.83
MFCD00008409
JBWYRBLDOOOJEU-UHFFFAOYSA-N
4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
96831
1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 127°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C(C6H4OCH3)2Cl
Quantity 25 g
Fieser 01,188
Formula Weight 338.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 255-002-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit