Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-1,3-cyclohexanedione, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
562-46-9
C8H12O2
140.18
MFCD00075224
PLGPBTCNKJQJHQ-UHFFFAOYSA-N
4,4-dimethyl-1,3-cyclohexanedione,1,3-cyclohexanedione, 4,4-dimethyl,4,4-dimethyl-cyclohexane-1,3-dione,acmc-209lre,4,4-dimethyl-1,3cyclohexanedione,4,4dimethyl-1,3-cyclohexanedione,4,4-dimethy 1,3-cyclohexanedione,1,3-cyclohexanedione,4,4-dimethyl,4,4-dimethyl-1,3-cyclohexanedione #
136360
4,4-dimethylcyclohexane-1,3-dione
CC1(CCC(=O)CC1=O)C
562-46-9
C8H12O2
140.18
MFCD00075224
PLGPBTCNKJQJHQ-UHFFFAOYSA-N
4,4-dimethyl-1,3-cyclohexanedione,1,3-cyclohexanedione, 4,4-dimethyl,4,4-dimethyl-cyclohexane-1,3-dione,acmc-209lre,4,4-dimethyl-1,3cyclohexanedione,4,4dimethyl-1,3-cyclohexanedione,4,4-dimethy 1,3-cyclohexanedione,1,3-cyclohexanedione,4,4-dimethyl,4,4-dimethyl-1,3-cyclohexanedione #
136360
4,4-dimethylcyclohexane-1,3-dione
CC1(CCC(=O)CC1=O)C
Boiling Point 130°C (7.0mmHg)
Melting Point 102°C to 105°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents
Formula Weight 140.18
Physical Form Needles
Percent Purity 98%
Chemical Name or Material 4, 4-Dimethyl-1, 3-cyclohexanedione
Show More Show Less