Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4,4'-Dimethyl-2,2'-bipyridine, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4,4'-Dimethyl-2,2'-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethyl-2,2′-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.
Solubility
Soluble in ethanol, acetic acid, benzene, toluene. Highly soluble in water at pH < 2.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
4,4′-Dimethyl-2,2′-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.
Solubility
Soluble in ethanol, acetic acid, benzene, toluene. Highly soluble in water at pH < 2.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 171°C to 175°C |
| Quantity | 5 g |
| Beilstein | 128995 |
| Solubility Information | Soluble in ethanol,acetic acid,benzene,toluene. Highly soluble in water at pH < 2. |
| Formula Weight | 184.24 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4'-Dimethyl-2,2'-bipyridine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 214-483-2 |
| RTECSNumber | DW1765000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction