Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethyl-2,2'-bipyridyl, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1134-35-6
C12H12N2
184.24
MFCD00006441
NBPGPQJFYXNFKN-UHFFFAOYSA-N
4,4'-dimethyl-2,2'-bipyridine,4,4'-dimethyl-2,2'-bipyridyl,4,4'-dimethyl-2,2'-dipyridyl,2,2'-bi-4-picoline,2,2'-bipyridine, 4,4'-dimethyl,2,2'-bi gamma-picoline,2,2'-di-4-picolyl,4-methyl-2-4-methylpyridin-2-yl pyridine,2,2'-bipyridyl, 4,4'-dimethyl,4,4-dimethyl-2,2-bipyridyl
14338
4-methyl-2-(4-methylpyridin-2-yl)pyridine
CC1=CC(=NC=C1)C2=NC=CC(=C2)C
1134-35-6
C12H12N2
184.24
MFCD00006441
NBPGPQJFYXNFKN-UHFFFAOYSA-N
4,4'-dimethyl-2,2'-bipyridine,4,4'-dimethyl-2,2'-bipyridyl,4,4'-dimethyl-2,2'-dipyridyl,2,2'-bi-4-picoline,2,2'-bipyridine, 4,4'-dimethyl,2,2'-bi gamma-picoline,2,2'-di-4-picolyl,4-methyl-2-4-methylpyridin-2-yl pyridine,2,2'-bipyridyl, 4,4'-dimethyl,4,4-dimethyl-2,2-bipyridyl
14338
4-methyl-2-(4-methylpyridin-2-yl)pyridine
CC1=CC(=NC=C1)C2=NC=CC(=C2)C
Melting Point 174°C to 176°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 25g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Formula Weight 184.24
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material 4, 4'-Dimethyl-2, 2'-bipyridyl
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-483-2