Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-2-oxazoline, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
30093-99-3
C5H9NO
99.13
KOAMXHRRVFDWRQ-UHFFFAOYSA-N
4,4-dimethyl-2-oxazoline,4,4-dimethyloxazoline,4,4-dimethyl-4,5-dihydrooxazole,oxazole, 4,5-dihydro-4,4-dimethyl,4,5-dihydro-4,4-dimethyloxazole,pubchem19647,acmc-1cjzq,koamxhrrvfdwrq-uhfffaoysa,4,4-dimethyl-4,5-dihydro-1,3-oxazole,4,4-dimethyl-5h-1,3-oxazol-5-yl
121630
4,4-dimethyl-5H-1,3-oxazole
CC1(COC=N1)C
30093-99-3
C5H9NO
99.13
KOAMXHRRVFDWRQ-UHFFFAOYSA-N
4,4-dimethyl-2-oxazoline,4,4-dimethyloxazoline,4,4-dimethyl-4,5-dihydrooxazole,oxazole, 4,5-dihydro-4,4-dimethyl,4,5-dihydro-4,4-dimethyloxazole,pubchem19647,acmc-1cjzq,koamxhrrvfdwrq-uhfffaoysa,4,4-dimethyl-4,5-dihydro-1,3-oxazole,4,4-dimethyl-5h-1,3-oxazol-5-yl
121630
4,4-dimethyl-5H-1,3-oxazole
CC1(COC=N1)C
Specific Gravity 0.94
Density 0.9400g/mL
Color Colorless
Infrared Spectrum Authentic
Flash Point 1°C
Refractive Index 1.4190 to 1.4240
Quantity 50g
Boiling Point 99.0°C to 100.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 99.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4, 4-Dimethyl-2-oxazoline
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

EINECSNumber : 250-045-7