Alfa Aesar™ 4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester, 98+%

Manufacturer: Alfa Aesar™ H54402MB

Quantity 25mg

 View more versions of this product

Catalog No. AAH54402MB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 146998-31-4
    Molecular Formula C19H18N4O4
    Formula Weight 366.37
    MDL Number MFCD00467415
    Synonym DABCYL-N-succinimidyl ester; 4-[(Dimethylamino)azo]benzene-4'-carboxylic acid N-succinimidyl ester
    Chemical Name or Material 4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester

    Specifications

    Formula Weight 366.37
    Molecular Formula C19H18N4O4
    MDL Number MFCD00467415
    CAS 146998-31-4
    Quantity 25mg
    Synonym DABCYL-N-succinimidyl ester; 4-[(Dimethylamino)azo]benzene-4'-carboxylic acid N-succinimidyl ester
    Recommended Storage −30° to −10°C
    Chemical Name or Material 4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester
    Assay ≥98%

    Description

    Applications
    4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester is a useful reagent (similar to MANCYL-SE) for labeling proteins or peptides through their amino-groups by forming stable peptide bonds. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles for fluorescent studies of proteins. Its fluorescent properties (has a characteristic broad and intense visible absorption but has no fluorescence) make it an ideal long wavelength quencher and it has been utilized as an acceptor chromophore in FRET studies. It is also used for N-terminal modification of peptides in automated synthesis.

    Solubility
    Soluble in DMSO.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents and heat.

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit