Alfa Aesar™ 4,4-Dimethylcyclohexanone, 98%

Manufacturer: Alfa Aesar™ H25857MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25857MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4255-62-3
    Molecular Formula C8H14O
    Formula Weight 126.2
    MDL Number MFCD00234965
    Chemical Name or Material 4,4-Dimethylcyclohexanone

    Specifications

    Formula Weight 126.2
    Molecular Formula C8H14O
    MDL Number MFCD00234965
    CAS 4255-62-3
    Flash Point 62°C (144°F)
    Melting Point 41° to 43°C
    Quantity 250mg
    Chemical Name or Material 4,4-Dimethylcyclohexanone
    Assay 98%

    Description

    Applications
    4,4-Dimethylcyclohexanone is used as a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit