Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4''-Dinitro-p-terphenyl, 98+%

Click to view available options
Quantity:
5g
25g
3282-11-9
C8H7NOS
165.21
MFCD00051743
QKAOOWJWWKWWOZ-UHFFFAOYSA-N
2-methoxyphenyl isothiocyanate,benzene, 1-isothiocyanato-2-methoxy,o-methoxyphenyl isothiocyanate,2-methoxyphenylisothiocyanate,1-isothiocyanato-2-methoxy-benzene,isothiocyanic acid 2-methoxyphenyl ester,2-methoxybenzenisothiocyanate,acmc-1coqu,o-methoxyphenylisothiocyanat,orthomethoxyphenyl isothiocyanate
520599
1-isothiocyanato-2-methoxybenzene
COC1=CC=CC=C1N=C=S
3282-11-9
C8H7NOS
165.21
MFCD00051743
QKAOOWJWWKWWOZ-UHFFFAOYSA-N
2-methoxyphenyl isothiocyanate,benzene, 1-isothiocyanato-2-methoxy,o-methoxyphenyl isothiocyanate,2-methoxyphenylisothiocyanate,1-isothiocyanato-2-methoxy-benzene,isothiocyanic acid 2-methoxyphenyl ester,2-methoxybenzenisothiocyanate,acmc-1coqu,o-methoxyphenylisothiocyanat,orthomethoxyphenyl isothiocyanate
520599
1-isothiocyanato-2-methoxybenzene
COC1=CC=CC=C1N=C=S
Quantity 5g
Melting Point 276°C to 280°C
Beilstein 2160043
Formula Weight 320.31
Percent Purity ≥98%
Chemical Name or Material 4,4''-Dinitro-p-terphenyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures