Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4''-Dinitro-p-terphenyl, 98+%

Click to view available options
Quantity:
5g
25g
3282-11-9
C18H12N2O4
320.30
MFCD00051743
MHOAYDHUNGLDTB-UHFFFAOYSA-N
4,4-dinitro-p-terphenyl,p-terphenyl, 4,4-dinitro,4,4-dinitro-1,1',4',1 terphenyl,1,4-bis 4-nitrophenyl benzene,p,p-dinitro-p-terphenyl,4,4-dinitro-para-terphenyl,4,4-dinitro-1,1',4',1-terphenyl,4,4-dinitro-1,1':4',1-terphenyl,1-nitro-4-4-4-nitrophenyl phenyl benzene,4,4-dinitro-1,1':4',1-terbenzene
101423
1,4-bis(4-nitrophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-]
3282-11-9
C18H12N2O4
320.30
MFCD00051743
MHOAYDHUNGLDTB-UHFFFAOYSA-N
4,4-dinitro-p-terphenyl,p-terphenyl, 4,4-dinitro,4,4-dinitro-1,1',4',1 terphenyl,1,4-bis 4-nitrophenyl benzene,p,p-dinitro-p-terphenyl,4,4-dinitro-para-terphenyl,4,4-dinitro-1,1',4',1-terphenyl,4,4-dinitro-1,1':4',1-terphenyl,1-nitro-4-4-4-nitrophenyl phenyl benzene,4,4-dinitro-1,1':4',1-terbenzene
101423
1,4-bis(4-nitrophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-]
Quantity 25g
Melting Point 276°C to 280°C
Beilstein 2160043
Formula Weight 320.31
Percent Purity ≥98%
Chemical Name or Material 4,4''-Dinitro-p-terphenyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning