Alfa Aesar™ 4,4-Diphenyl-2-butanone, 98%

Manufacturer: Alfa Aesar™ A1384703

Quantity 1g

 View more versions of this product

Catalog No. AAA1384703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5409-60-9
    Molecular Formula C16H16O
    Formula Weight 224.3
    MDL Number MFCD00017619
    Chemical Name or Material 4,4-Diphenyl-2-butanone

    Specifications

    Formula Weight 224.3
    Molecular Formula C16H16O
    MDL Number MFCD00017619
    Quantity 1g
    CAS 5409-60-9
    Chemical Name or Material 4,4-Diphenyl-2-butanone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit