Alfa Aesar™ 4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%

Manufacturer: Alfa Aesar™ B2314214

Quantity 25g

 View more versions of this product

Catalog No. AAB2314214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25519-78-2
    Molecular Formula C12H14FNO·HCl
    Formula Weight 243.71
    MDL Number MFCD00044912
    Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride

    Specifications

    Formula Weight 243.71
    Molecular Formula C12H14FNO·HCl
    MDL Number MFCD00044912
    CAS 25519-78-2
    Quantity 25g
    Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit