Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
25519-78-2
C12H15ClFNO
243.706
MFCD00044912
GPKDBZQZPNOBGM-UHFFFAOYSA-N
4-4-fluorobenzoyl piperidine hydrochloride,4-fluorophenyl piperidin-4-yl methanone hydrochloride,4-fluorophenyl 4-piperidinyl ketone hydrochloride,p-fluorophenyl 4-piperidyl ketone hydrochloride,4-fluorophenyl 4-piperidyl methanone hydrochloride,4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride,4-4-fluorobenzoyl piperidine hcl,4-p-fluorobenzoyl piperidine hydrochloride,4-4'-fluorobenzoyl piperidine hydrochloride,methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
3084438
(4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
C1CNCCC1C(=O)C2=CC=C(C=C2)F.Cl
25519-78-2
C12H15ClFNO
243.706
MFCD00044912
GPKDBZQZPNOBGM-UHFFFAOYSA-N
4-4-fluorobenzoyl piperidine hydrochloride,4-fluorophenyl piperidin-4-yl methanone hydrochloride,4-fluorophenyl 4-piperidinyl ketone hydrochloride,p-fluorophenyl 4-piperidyl ketone hydrochloride,4-fluorophenyl 4-piperidyl methanone hydrochloride,4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride,4-4-fluorobenzoyl piperidine hcl,4-p-fluorobenzoyl piperidine hydrochloride,4-4'-fluorobenzoyl piperidine hydrochloride,methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
3084438
(4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
C1CNCCC1C(=O)C2=CC=C(C=C2)F.Cl
Quantity 5g
Formula Weight 243.71
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride