Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Fluorobenzyloxy)benzaldehyde, 97%

Click to view available options
Quantity:
5g
25g
56442-17-2
C14H11FO2
230.238
MFCD00173650
QGULWBQOCMQNFD-UHFFFAOYSA-N
4-4-fluorobenzyloxy benzaldehyde,4-4-fluorobenzyl oxy benzaldehyde,4-4-fluorophenyl methoxy benzaldehyde,acmc-1b0gw,cbmicro_008348,4-4-fluorobenzyl-oxy benzaldehyde,4-4-fluoro-benzyloxy-benzaldehyde,benzaldehyde,4-4-fluorophenyl methoxy
735860
4-[(4-fluorophenyl)methoxy]benzaldehyde
C1=CC(=CC=C1COC2=CC=C(C=C2)C=O)F
56442-17-2
C14H11FO2
230.238
MFCD00173650
QGULWBQOCMQNFD-UHFFFAOYSA-N
4-4-fluorobenzyloxy benzaldehyde,4-4-fluorobenzyl oxy benzaldehyde,4-4-fluorophenyl methoxy benzaldehyde,acmc-1b0gw,cbmicro_008348,4-4-fluorobenzyl-oxy benzaldehyde,4-4-fluoro-benzyloxy-benzaldehyde,benzaldehyde,4-4-fluorophenyl methoxy
735860
4-[(4-fluorophenyl)methoxy]benzaldehyde
C1=CC(=CC=C1COC2=CC=C(C=C2)C=O)F
Quantity 25g
Sensitivity Air Sensitive
Formula Weight 230.24
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorobenzyloxy)benzaldehyde

TSCA : No

Recommended Storage : Ambient temperatures