Alfa Aesar™ 4-(4-Fluorophenoxy)benzonitrile, 98%

Manufacturer: Alfa Aesar™ H3253014

Quantity 25g

 View more versions of this product

Catalog No. AAH3253014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 215589-24-5
    Molecular Formula C13H8FNO
    Formula Weight 213.21
    MDL Number MFCD01815090
    Chemical Name or Material 4-(4-Fluorophenoxy)benzonitrile

    Specifications

    Formula Weight 213.21
    Molecular Formula C13H8FNO
    MDL Number MFCD01815090
    CAS 215589-24-5
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 67° to 68°C
    Quantity 25g
    Chemical Name or Material 4-(4-Fluorophenoxy)benzonitrile
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit