Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Hydroxyphenyl)-2-butanone, 99+%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
5471-51-2
C10H12O2
164.2
MFCD00002394
NJGBTKGETPDVIK-UHFFFAOYSA-N
4-4-hydroxyphenyl-2-butanone,raspberry ketone,4-4-hydroxyphenyl butan-2-one,frambinone,oxyphenalon,rheosmin,4-p-hydroxyphenyl-2-butanone,4-hydroxybenzylacetone,betuligenol,rasketone
21648
CHEBI:68656
4-(4-hydroxyphenyl)butan-2-one
CC(=O)CCC1=CC=C(C=C1)O
5471-51-2
C10H12O2
164.2
MFCD00002394
NJGBTKGETPDVIK-UHFFFAOYSA-N
4-4-hydroxyphenyl-2-butanone,raspberry ketone,4-4-hydroxyphenyl butan-2-one,frambinone,oxyphenalon,rheosmin,4-p-hydroxyphenyl-2-butanone,4-hydroxybenzylacetone,betuligenol,rasketone
21648
CHEBI:68656
4-(4-hydroxyphenyl)butan-2-one
CC(=O)CCC1=CC=C(C=C1)O
Melting Point 82°C to 84°C
Packaging Glass bottle
Quantity 250g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula HOC6H4CH2CH2COCH3
Formula Weight 164.2
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-806-4

RTECSNumber : EL8925000

TSCA : TSCA