Alfa Aesar™ 4-(4-Methoxyphenyl)-2-butanone, 99%

Manufacturer: Alfa Aesar™ A1101109

Quantity 10g

 View more versions of this product

Catalog No. AAA1101109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-20-1
    Molecular Formula C11H14O2
    Formula Weight 178.23
    MDL Number MFCD00008791
    Beilstein 1869592
    Synonym p-Anisylacetone
    Chemical Name or Material 4- (4-Methoxyphenyl)-2-butanone

    Specifications

    Formula Weight 178.23
    Boiling Point 147° to 148°C (8mmHg)
    Beilstein 1869592
    Synonym p-Anisylacetone
    Quantity 10g
    RTECS Number EL9085500
    TSCA TSCA
    Molecular Formula C11H14O2
    CAS 104-20-1
    Melting Point 7° to 9°C
    MDL Number MFCD00008791
    Flash Point >110°C (230°F)
    Chemical Name or Material 4- (4-Methoxyphenyl)-2-butanone
    Assay 99%

    Description

    Applications
    4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.

    Solubility
    Not miscible in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit