Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Methoxyphenyl)-2-methylthiazole, 97%

Click to view available options
Quantity:
1g
5g
25g
50834-78-1
C11H11NOS
205.275
MFCD00460502
OVKCVKLSCKKRJY-UHFFFAOYSA-N
4-4-methoxyphenyl-2-methylthiazole,4-4-methoxy-phenyl-2-methyl-thiazole,4-4-methoxyphenyl-2-methyl-1,3-thiazole,thiazole, 4-4-methoxyphenyl-2-methyl,acmc-1ampl,2-methyl-4-4-methoxyphenyl thiazole,4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene
768765
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
CC1=NC(=CS1)C2=CC=C(C=C2)OC
50834-78-1
C11H11NOS
205.275
MFCD00460502
OVKCVKLSCKKRJY-UHFFFAOYSA-N
4-4-methoxyphenyl-2-methylthiazole,4-4-methoxy-phenyl-2-methyl-thiazole,4-4-methoxyphenyl-2-methyl-1,3-thiazole,thiazole, 4-4-methoxyphenyl-2-methyl,acmc-1ampl,2-methyl-4-4-methoxyphenyl thiazole,4-methoxy-1-2-methyl 1,3-thiazol-4-yl benzene
768765
4-(4-methoxyphenyl)-2-methyl-1,3-thiazole
CC1=NC(=CS1)C2=CC=C(C=C2)OC
Quantity 5g
Color Cream
Melting Point 64°C to 69°C
Formula Weight 205.28
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-(4-Methoxyphenyl)-2-methylthiazole