Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methoxyphenyl)butyric acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4521-28-2
C11H14O3
194.23
MFCD00004404
LZHMNCJMXQKSBY-UHFFFAOYSA-N
4-4-methoxyphenyl butyric acid,4-4-methoxyphenyl butanoic acid,benzenebutanoic acid, 4-methoxy,4-4-methoxyphenyl butyricacid,4-4-methoxyphenyl-1-butyric acid,p-methoxy-g-phenylbutyric acid,pubchem15207,4-methoxybenzenebutyric acid,4-p-methoxyphenylbutyric acid
78280
4-(4-methoxyphenyl)butanoic acid
COC1=CC=C(C=C1)CCCC(=O)O
4521-28-2
C11H14O3
194.23
MFCD00004404
LZHMNCJMXQKSBY-UHFFFAOYSA-N
4-4-methoxyphenyl butyric acid,4-4-methoxyphenyl butanoic acid,benzenebutanoic acid, 4-methoxy,4-4-methoxyphenyl butyricacid,4-4-methoxyphenyl-1-butyric acid,p-methoxy-g-phenylbutyric acid,pubchem15207,4-methoxybenzenebutyric acid,4-p-methoxyphenylbutyric acid
78280
4-(4-methoxyphenyl)butanoic acid
COC1=CC=C(C=C1)CCCC(=O)O
Infrared Spectrum Authentic
Melting Point 56.0°C to 61.0°C
Quantity 25g
Color Cream to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4(CH2)3CO2H
Beilstein 10,IV,683
Formula Weight 194.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-(4-Methoxyphenyl)butyric acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-849-3