Alfa Aesar™ 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

Manufacturer: Alfa Aesar™ H5111603

Quantity 1g

 View more versions of this product

Catalog No. AAH5111603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 871366-38-0
    Molecular Formula C20H24BNO3
    Formula Weight 337.22
    MDL Number MFCD10567027
    Synonym 1-Methoxyphenyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
    Chemical Name or Material 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 337.22
    Molecular Formula C20H24BNO3
    MDL Number MFCD10567027
    CAS 871366-38-0
    Synonym 1-Methoxyphenyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
    Quantity 1g
    Chemical Name or Material 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit