Alfa Aesar™ 4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 96%

Manufacturer: Alfa Aesar™ H5165903

Quantity 1g

 View more versions of this product

Catalog No. AAH5165903

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 747413-21-4
    Molecular Formula C17H27BN2O2
    Formula Weight 302.22
    MDL Number MFCD06795650
    Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
    Chemical Name or Material 4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 302.22
    Molecular Formula C17H27BN2O2
    MDL Number MFCD06795650
    CAS 747413-21-4
    Synonym 1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
    Quantity 1g
    Chemical Name or Material 4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester
    Assay 96%

    Description

    Applications
    4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit