Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4′-Methylenebis(N,N-dimethylaniline), 98%, Acros Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
101-61-1
C17H22N2
254.37
MFCD00008317
JNRLEMMIVRBKJE-UHFFFAOYSA-N
4,4'-methylenebis n,n-dimethylaniline,tetra-base,tetrabase,michler's base,methane base,methylene base,michler's hydride,michler's methane,4,4'-bis dimethylamino diphenylmethane,reduced michler's ketone
7567
CHEBI:34370
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C
101-61-1
C17H22N2
254.37
MFCD00008317
JNRLEMMIVRBKJE-UHFFFAOYSA-N
4,4'-methylenebis n,n-dimethylaniline,tetra-base,tetrabase,michler's base,methane base,methylene base,michler's hydride,michler's methane,4,4'-bis dimethylamino diphenylmethane,reduced michler's ketone
7567
CHEBI:34370
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C
Color Beige-Brown or Gray or White
Infrared Spectrum Authentic
Flash Point 178°C
Packaging Glass bottle
Boiling Point 390.0°C
Melting Point 86.0°C to 90.0°C
Quantity 500g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H4CH2C6H4NH2
Beilstein 13, 239
Merck Index 15, 6255
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,ether,carbon disulfide and,acids,slightly soluble in cold alcohol,more soluble in,hot alcohol
Formula Weight 254.37
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 4, 4'-Methylenebis(N, N-dimethylaniline), 99+%
Show More Show Less

GHS H Statement:
May cause cancer.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-959-2

RTECSNumber : BY5250000

TSCA : TSCA