Alfa Aesar™ 4-(4-Morpholinyl)benzoic acid, 97%

Manufacturer: Alfa Aesar™ H3225806

Quantity 5g

 View more versions of this product

Catalog No. AAH3225806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7470-38-4
    Molecular Formula C11H13NO3
    Formula Weight 207.23
    MDL Number MFCD00588379
    Synonym 4-Morpholinobenzoic acid
    Chemical Name or Material 4-(4-Morpholinyl)benzoic acid

    Specifications

    Formula Weight 207.23
    MDL Number MFCD00588379
    Melting Point 285° to 288°C
    Recommended Storage 2° to 8°C
    Molecular Formula C11H13NO3
    CAS 7470-38-4
    Synonym 4-Morpholinobenzoic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Chemical Name or Material 4-(4-Morpholinyl)benzoic acid
    Assay 97%

    Description

    Applications
    4-(4-Morpholinyl)benzoic acid, is used as an intermediate in organic synthesis. The water based additive is used in a variety of applications as a heat-sealing and anti-blocking agent.

    Solubility
    Slightly soluble in water.

    Notes
    Moisture & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, water and heat.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit