Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Nitrobenzyl)pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
1083-48-3
C12H10N2O2
214.22
MFCD00006445
MNHKUCBXXMFQDM-UHFFFAOYSA-N
4-4-nitrobenzyl pyridine,4-4-nitrophenyl methyl pyridine,pyridine, 4-4-nitrophenyl methyl,pyridine, 4-p-nitrobenzyl,1-p-nitrobenzyl pyridine,4-p-nitrobenzyl pyridine,4 4-nitrobenzyl primide,unii-0z6gs37n7h,gamma-p-nitrobenzyl pyridine
14129
4-[(4-nitrophenyl)methyl]pyridine
C1=CC(=CC=C1CC2=CC=NC=C2)[N+](=O)[O-]
1083-48-3
C12H10N2O2
214.22
MFCD00006445
MNHKUCBXXMFQDM-UHFFFAOYSA-N
4-4-nitrobenzyl pyridine,4-4-nitrophenyl methyl pyridine,pyridine, 4-4-nitrophenyl methyl,pyridine, 4-p-nitrobenzyl,1-p-nitrobenzyl pyridine,4-p-nitrobenzyl pyridine,4 4-nitrobenzyl primide,unii-0z6gs37n7h,gamma-p-nitrobenzyl pyridine
14129
4-[(4-nitrophenyl)methyl]pyridine
C1=CC(=CC=C1CC2=CC=NC=C2)[N+](=O)[O-]
Melting Point 69°C to 72°C
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)CH2C6H4NO2
Beilstein 20, II, 272
Solubility Information Solubility in water: 1,04g/L
Formula Weight 214.22
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-(4-Nitrobenzyl)pyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning