Alfa Aesar™ 4'-(4-Nitrobenzyloxy)acetophenone, 97%

Manufacturer: Alfa Aesar™ A1673806

Quantity 5g

 View more versions of this product

Catalog No. AAA1673806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 205748-03-4
    Molecular Formula C15H13NO4
    Formula Weight 271.28
    MDL Number MFCD00075690
    Beilstein 8321694
    Chemical Name or Material 4'-(4-Nitrobenzyloxy)acetophenone

    Specifications

    Formula Weight 271.28
    Molecular Formula C15H13NO4
    MDL Number MFCD00075690
    Beilstein 8321694
    Melting Point 129° to 131°C
    Quantity 5g
    CAS 205748-03-4
    Chemical Name or Material 4'-(4-Nitrobenzyloxy)acetophenone
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit