Alfa Aesar™ 4-(4-Nitrobenzyloxy)benzaldehyde, 98%

Manufacturer: Alfa Aesar™ A1106706

Quantity 5g

 View more versions of this product

Catalog No. AAA1106706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67565-48-4
    Molecular Formula C14H11NO4
    Formula Weight 257.25
    MDL Number MFCD00017041
    Beilstein 1884256
    Chemical Name or Material 4- (4-Nitrobenzyloxy)benzaldehyde

    Specifications

    Formula Weight 257.25
    Molecular Formula C14H11NO4
    Melting Point 128° to 132°C
    Quantity 5g
    MDL Number MFCD00017041
    CAS 67565-48-4
    Beilstein 1884256
    Chemical Name or Material 4- (4-Nitrobenzyloxy)benzaldehyde
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit