Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(4-Nitrophenoxy)benzoic acid, 97%

Manufacturer:  Alfa Aesar™ H3279406

Quantity 5g

 View more versions of this product

Catalog No. AAH3279406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16309-45-8
259.22
UN3077
C13H9NO5
MFCD00429494
4-(4-Nitrophenoxy)benzoic acid

Specifications

238° to 242°C
16309-45-8
259.22
UN3077
97%
5g
C13H9NO5
MFCD00429494
4-(4-Nitrophenoxy)benzoic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit