Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Nitrophenylazo)catechol, 98%, Thermo Scientific™

Catalog Number AAB2408606
Click to view available options
Quantity:
5g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 843-33-4
Odor Odorless
Molecular Formula C12H9N3O4
Quantity 5g
MDL Number MFCD00014711
Synonym 4-4-nitrophenylazo catechol, 4-4-nitrobenzeneazo catechol, 4-e-2-4-nitrophenyl diazen-1-yl benzene-1,2-diol, 4-2-4-nitrophenyl diazen-1-yl benzene-1,2-diol, 4-4-nitrophenyl azo pyrocatechol, acmc-209pua, 4'-nitroazobenzene-3,4-diol, 4-p-nitrophenylazo-catechol, 3,4-dihydroxy-4'-nitroazobenzol, dgozwusvobkxnr-uhfffaoysa-n
InChI Key MKFXYEBJVGGEKE-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC=C(NNC2=CC(=O)C(=O)C=C2)C=C1
IUPAC Name 4-[2-(4-nitrophenyl)hydrazin-1-yl]cyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol) 259.22
PubChem CID 5357361
Formula Weight 259.22
Percent Purity 98%
Chemical Name or Material 4-(4-Nitrophenylazo)catechol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-676-6
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit