Alfa Aesar™ 4-(4-Nitrophenylazo)resorcinol, 95%

Manufacturer: Alfa Aesar™ A1223222

Quantity 100g

 View more versions of this product

Catalog No. AAA1223222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74-39-5
    Molecular Formula C12H9N3O4
    Formula Weight 259.22
    MDL Number MFCD00007310
    Beilstein 674709
    Merck Index 14,5695
    Synonym 2,4-Dihydroxy-4'-nitroazobenzene; Magneson
    Chemical Name or Material 4- (4-Nitrophenylazo)resorcinol

    Specifications

    Formula Weight 259.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C12H9N3O4
    RTECS Number VH2810000
    Melting Point ≈185°C (dec.)
    TSCA TSCA
    Synonym 2,4-Dihydroxy-4'-nitroazobenzene; Magneson
    Health Hazard 1 Warning
    MDL Number MFCD00007310
    Beilstein 674709
    Merck Index 14,5695
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 100g
    CAS 74-39-5
    Chemical Name or Material 4- (4-Nitrophenylazo)resorcinol
    Assay 95%

    Description

    Applications
    It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.

    Solubility
    Soluble in dilute sodium hydroxide. Insoluble in water.

    Notes
    Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit