Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Oxybis(benzoic acid), 98+%

Click to view available options
Quantity:
25g
100g
2215-89-6
C14H10O5
258.229
MFCD00013988
WVDRSXGPQWNUBN-UHFFFAOYSA-N
4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
75183
4-(4-carboxyphenoxy)benzoic acid
C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O
2215-89-6
C14H10O5
258.229
MFCD00013988
WVDRSXGPQWNUBN-UHFFFAOYSA-N
4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
75183
4-(4-carboxyphenoxy)benzoic acid
C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O
Quantity 25g
Melting Point 329°C to 332°C
Beilstein 2218315
Formula Weight 258.23
Percent Purity ≥98%
Chemical Name or Material 4,4'-Oxybis(benzoic acid)

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-683-0

TSCA : Yes

Recommended Storage : Ambient temperatures