Alfa Aesar™ 4-(4-Pyridyl)benzoic acid, 97%

Manufacturer: Alfa Aesar™ H2788206

Quantity 5g

 View more versions of this product

Catalog No. AAH2788206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4385-76-6
    Molecular Formula C12H9NO2
    Formula Weight 199.2
    MDL Number MFCD04114574
    Chemical Name or Material 4-(4-Pyridyl)benzoic acid

    Specifications

    Formula Weight 199.2
    Molecular Formula C12H9NO2
    MDL Number MFCD04114574
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point >300°C
    Quantity 5g
    CAS 4385-76-6
    Chemical Name or Material 4-(4-Pyridyl)benzoic acid
    Assay 97%

    Description

    Applications
    Self-assembly of 4-(4-pyridyl)benzoic acid (pybenH), Ni2+, SCN- and an aromatic guest (naphthalene or dibenz[a,h]anthracene) gave two isostructural inclusion compounds, in which pybenH ligands form the corresponding dimers acting as bidentate building blocks with a length of ca. 20.86 Å. By replacing an isonicotinic acid dimer with a 4-(4-pyridyl)benzoic acid (pybenH) dimer, new inclusion compounds in which pybenH dimers function as bridging ligands of ca. 21 Å.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit