Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4′-Thiobis(6-tert-butyl-o-cresol) 98.0+%, TCI America™

Click to view available options
Quantity:
25g
96-66-2
C22H30O2S
358.54
MFCD00008823
YFHKLSPMRRWLKI-UHFFFAOYSA-N
Bis(3-tert-butyl-4-hydroxy-5-methylphenyl) Sulfide
7306
2-tert-butyl-4-(3-tert-butyl-4-hydroxy-5-methylphenyl)sulfanyl-6-methylphenol
CC1=CC(=CC(=C1O)C(C)(C)C)SC2=CC(=C(C(=C2)C)O)C(C)(C)C
96-66-2
C22H30O2S
358.54
MFCD00008823
YFHKLSPMRRWLKI-UHFFFAOYSA-N
Bis(3-tert-butyl-4-hydroxy-5-methylphenyl) Sulfide
7306
2-tert-butyl-4-(3-tert-butyl-4-hydroxy-5-methylphenyl)sulfanyl-6-methylphenol
CC1=CC(=CC(=C1O)C(C)(C)C)SC2=CC(=C(C(=C2)C)O)C(C)(C)C
Quantity 25g
Color Undesignated
Melting Point 127°C
Formula Weight 358.54
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4,4′-Thiobis(6-tert-butyl-o-cresol)

EINECSNumber : (3)-2903

RTECSNumber : GP3200000

TSCA : Yes