Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Thiodiphenol, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
2664-63-3
C12H10O2S
218.28
VWGKEVWFBOUAND-UHFFFAOYSA-N
4,4'-thiodiphenol,bis 4-hydroxyphenyl sulfide,thiobisphenol,4,4'-thiobis-phenol,phenol, 4,4'-thiobis,4,4'-thiobisphenol,4,4'-dihydroxydiphenyl sulfide,bis 4-oxyphenyl sulfide,p,p'-dihydroxydiphenyl sulfide,4,4'-dioxydiphenyl sulfide
17570
CHEBI:38957
4-(4-hydroxyphenyl)sulfanylphenol
C1=CC(=CC=C1O)SC2=CC=C(C=C2)O
2664-63-3
C12H10O2S
218.28
VWGKEVWFBOUAND-UHFFFAOYSA-N
4,4'-thiodiphenol,bis 4-hydroxyphenyl sulfide,thiobisphenol,4,4'-thiobis-phenol,phenol, 4,4'-thiobis,4,4'-thiobisphenol,4,4'-dihydroxydiphenyl sulfide,bis 4-oxyphenyl sulfide,p,p'-dihydroxydiphenyl sulfide,4,4'-dioxydiphenyl sulfide
17570
CHEBI:38957
4-(4-hydroxyphenyl)sulfanylphenol
C1=CC(=CC=C1O)SC2=CC=C(C=C2)O
Color Brown to White
Melting Point 151.0°C to 154.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula S(C6H4OH)2
Beilstein 06,86
Formula Weight 218.28
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4, 4'-Thiodiphenol
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

GHS Signal Word: Danger

EINECSNumber : 220-197-9